accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175204-53-2

IUPAC Name: 4-(methylsulfanyl)-6-(pyridin-2-yl)-1,3,5-triazin-2-amine
Synonyms: 4-(methylsulfanyl)-6-(pyridin-2-yl)-1,3,5-triazin-2-amine
Molecular Weight (g/mol): 219.27
Molecular Formula: C9H9N5S
InChi Key: MAFCMTDUOYKYOR-UHFFFAOYSA-N
SMILES: CSC1=NC(=NC(N)=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results