accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175204-82-7

IUPAC Name: methyl 4-(trifluoromethyl)pyridine-3-carboxylate
Synonyms: methyl 4-(trifluoromethyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: RFUABQNZKSSKRX-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CN=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results