accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONHSHN

CAS RN 175205-07-9

IUPAC Name: 6-(methoxymethyl)-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: 6-(methoxymethyl)-2-sulfanylidene-1,3-dihydropyrimidin-4-one
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: YNVMVIUSRGXSEN-UHFFFAOYSA-N
SMILES: COCC1=CC(=O)NC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results