accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNNHCl

CAS RN 175205-49-9

IUPAC Name: 1-[4-(1,2,3-thiadiazol-4-yl)phenyl]methanamine hydrochloride
Synonyms: 1-[4-(1,2,3-thiadiazol-4-yl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 227.71
Molecular Formula: C9H10ClN3S
InChi Key: WUUGGOQPXZDQNW-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(C=C1)C1=CSN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results