accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175205-62-6

IUPAC Name: 3-(chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Synonyms: 3-(chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 224.64
Molecular Formula: C10H9ClN2O2
InChi Key: GSWMZQIUADKXLN-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=NC(CCl)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results