CAS RN 175205-81-9
IUPAC Name:
2-bromo-4-(trifluoromethyl)pyridine
Synonyms:
2-bromo-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
226
Molecular Formula:
C6H3BrF3N
InChi Key:
WZVHLUMAQLUNTJ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Br)=NC=C1
1
–
2
of
2
results
1
2-Bromo-4-(trifluoromethyl)pyridine, 97%
CAS: 175205-81-9 Molecular Formula: C6H3BrF3N Molecular Weight (g/mol): 225.996 MDL Number: MFCD00153085 InChI Key: WZVHLUMAQLUNTJ-UHFFFAOYSA-N Synonym: 2-bromo-4-trifluoromethyl pyridine,pyridine, 2-bromo-4-trifluoromethyl,2-bromo-4-trifluoromethyl-pyridine,2-bromo-4-trifluormethyl pyridine,pubchem8064,acmc-209e9d,abbypharma ap-11-5529 PubChem CID: 2781543 IUPAC Name: 2-bromo-4-(trifluoromethyl)pyridine SMILES: C1=CN=C(C=C1C(F)(F)F)Br