accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175276-92-3

FFSH2N

CAS RN 175276-92-3

IUPAC Name: 2,4-difluorobenzene-1-carbothioamide
Synonyms: 2,4-difluorobenzenecarbothioamide
Molecular Weight (g/mol): 173.18
Molecular Formula: C7H5F2NS
InChi Key: MOHAZBCWHUCIEX-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results