accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175278-17-8

NH2OFFFBr

CAS RN 175278-17-8

IUPAC Name: 2-bromo-4-(trifluoromethoxy)aniline
Synonyms: 2-bromo-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 256.02
Molecular Formula: C7H5BrF3NO
InChi Key: ROSTYHNIIDIBEG-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(F)(F)F)C=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results