accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHOHNONOOH2O

CAS RN 175278-58-7

IUPAC Name: 5-nitro-1,3-diazinane-2,4,6-trione hydrate
Synonyms: 5-nitro-1,3-diazinane-2,4,6-trione hydrate
Molecular Weight (g/mol): 191.1
Molecular Formula: C4H5N3O6
InChi Key: YKGZAIJTCJOZNK-UHFFFAOYSA-N
SMILES: O.[O-][N+](=O)C1C(=O)NC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results