accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1753-75-9

IUPAC Name: 5-bromo-2,1,3-benzothiadiazole
Synonyms: 5-bromo-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 215.07
Molecular Formula: C6H3BrN2S
InChi Key: LLCRUZDFDGTAAN-UHFFFAOYSA-N
SMILES: BrC1=CC2=NSN=C2C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)