accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1757-42-2

IUPAC Name: 3-methylcyclopentan-1-one
Synonyms: 3-methylcyclopentan-1-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: AOKRXIIIYJGNNU-UHFFFAOYNA-N
SMILES: CC1CCC(=O)C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)