accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17579-99-6

IH3CPOOO

CAS RN 17579-99-6

IUPAC Name: methyltriphenoxyphosphanium iodide
Synonyms: methyltriphenoxyphosphanium iodide
Molecular Weight (g/mol): 452.23
Molecular Formula: C19H18IO3P
InChi Key: VKTOBGBZBCELGC-UHFFFAOYSA-M
SMILES: [I-].C[P+](OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results