accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17601-94-4

NH2BrNNOO

CAS RN 17601-94-4

IUPAC Name: 2-amino-3-bromo-5-nitrobenzonitrile
Synonyms: 2-amino-3-bromo-5-nitrobenzonitrile
Molecular Weight (g/mol): 242.03
Molecular Formula: C7H4BrN3O2
InChi Key: MUHLVSZIVTURCZ-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1C#N)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)