accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCSNOO

CAS RN 17614-74-3

IUPAC Name: 1-isothiocyanato-4-methyl-2-nitrobenzene
Synonyms: 1-isothiocyanato-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H6N2O2S
InChi Key: CUQUTWSUXWDJKF-UHFFFAOYSA-N
SMILES: CC1=CC=C(N=C=S)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results