accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4SN

CAS RN 1762-95-4

IUPAC Name: ammonium cyanosulfanide
Synonyms: ammonium thiocyanate
Molecular Weight (g/mol): 76.12
Molecular Formula: CH4N2S
InChi Key: SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES: [NH4+].[S-]C#N
Molecular Weight (g/mol) 
  • (24)
  • (14)
Quantity 
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
  • (7)
  • (3)
  • (13)
Grade 
  • (2)
  • (5)
  • (7)
  • (1)
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (4)
  • (7)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (15)
  • (6)
  • (5)
  • (2)
  • (3)
  • (24)
  • (14)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (13)
  • (2)
  • (5)
  • (7)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (4)
  • (7)
  • (2)
  • (2)