accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17625-83-1

NH2HNO

CAS RN 17625-83-1

IUPAC Name: N-(4-aminophenyl)benzamide
Synonyms: N-(4-aminophenyl)benzamide
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: GTTFJYUWPUKXJH-UHFFFAOYSA-N
SMILES: NC1=CC=C(NC(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results