accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(R)O(S)O(R)O(R)O(S)OHO(R)OH(S)OH(S)OH(R)OH(S)HO(S)HO(R)HO(S)HO(R)HO

CAS RN 17629-30-0

IUPAC Name: (2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol delta-(+)-raffinose raffinose, pure raffinose raffinose hydrate 6G-α-D-galactosylsucrose melitriose D-raffinose raffinose (8CI) 6G-α-delta-galactosylsucrose
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: MUPFEKGTMRGPLJ-ZQSKZDJDSA-N
SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Quantity 
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (9)
  • (3)
Physical Form 
  • (9)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (9)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)