accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CON

CAS RN 176445-77-5

IUPAC Name: 4-(dimethylamino)oxane-4-carbonitrile
Synonyms: 4-(dimethylamino)oxane-4-carbonitrile
Molecular Weight (g/mol): 154.21
Molecular Formula: C8H14N2O
InChi Key: FDGOISRHLNMQRE-UHFFFAOYSA-N
SMILES: CN(C)C1(CCOCC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results