accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CONH2

CAS RN 176445-80-0

IUPAC Name: 4-(aminomethyl)-N,N-dimethyloxan-4-amine
Synonyms: 4-(aminomethyl)-N,N-dimethyloxan-4-amine
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: KCLFWPREWYWLTP-UHFFFAOYSA-N
SMILES: CN(C)C1(CN)CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results