accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 176726-07-1

IUPAC Name: scandium(3+) tris(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: scandium(3+) tris(bistriflylimide anion)
Molecular Weight (g/mol): 885.36
Molecular Formula: C6F18N3O12S6Sc
InChi Key: FUXLYEZEIZAKTL-UHFFFAOYSA-N
SMILES: [Sc+3].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results