accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 176736-49-5

NOOOONOOOOH2OH2OH2OH2ONaNaZn

CAS RN 176736-49-5

IUPAC Name: zinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Synonyms: zinc(2+) disodium ion(4-) edta tetrahydrate
Molecular Weight (g/mol): 471.63
Molecular Formula: C10H20N2Na2O12Zn
InChi Key: ZEVLVCFKKCNROJ-UHFFFAOYSA-J
SMILES: O.O.O.O.[Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results