accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17696-69-4

IUPAC Name: dihydrogen 3-[20-(2-carboxyethyl)-9,14-bis(1-hydroxyethyl)-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoic acid dichloride
Synonyms: dihydrogen 3-[20-(2-carboxyethyl)-9,14-bis(1-hydroxyethyl)-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoic acid dichloride
Molecular Weight (g/mol): 671.62
Molecular Formula: C34H40Cl2N4O6
InChi Key: WZQQOXSHARXAID-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC(O)C1=C(C)C2=CC3=C(C(C)O)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results