accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONCS

CAS RN 17702-11-3

IUPAC Name: 1-isothiocyanato-3-methoxypropane
Synonyms: 1-isothiocyanato-3-methoxypropane
Molecular Weight (g/mol): 131.19
Molecular Formula: C5H9NOS
InChi Key: UKVFWFDXYBLOLU-UHFFFAOYSA-N
SMILES: COCCCN=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)