accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 17702-41-9

IUPAC Name: decaborane
Synonyms: decaborane
Molecular Weight (g/mol): 108.1
Molecular Formula: B10
InChi Key: KGUZAUWGHAIUGS-UHFFFAOYSA-N
SMILES: [B][B][B][B][B][B][B][B][B][B]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results