accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HC

CAS RN 17715-00-3

IUPAC Name: (prop-2-yn-1-yl)cyclohexane
Synonyms: prop-2-yn-1-ylcyclohexane
Molecular Weight (g/mol): 122.21
Molecular Formula: C9H14
InChi Key: UARFKZSJGDQRLF-UHFFFAOYSA-N
SMILES: C#CCC1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)