CAS RN 17720-18-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nicotinic Acid Riboside, TRC
CAS: 17720-18-2 Molecular Formula: C11 H13 N O6 Molecular Weight (g/mol): 255.22 Synonym: Nicotinic riboside,Nicotinic acid ribonucleoside IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]pyridin-1-ium-3-carboxylate SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)[n+]2cccc(c2)C(=O)[O-]