accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17737-65-4

CH3ClNHNOHO

CAS RN 17737-65-4

IUPAC Name: 2-[(3-chloro-2-methylphenyl)amino]pyridine-3-carboxylic acid
Synonyms: deltar clonix 氯尼辛 clonixinum clonixic acid clonixin clonixino clonixine
Molecular Weight (g/mol): 262.69
Molecular Formula: C13H11ClN2O2
InChi Key: CLOMYZFHNHFSIQ-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1NC1=NC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results