accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17740-40-8

CH3OONOOH3C

CAS RN 17740-40-8

IUPAC Name: 2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate
Synonyms: 2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate
Molecular Weight (g/mol): 305.37
Molecular Formula: C17H23NO4
InChi Key: LDUSEIANLSWKPY-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCC(N1CC1=CC=CC=C1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results