accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17783-69-6

H3CSOOO

CAS RN 17783-69-6

IUPAC Name: 2,3-dihydro-1H-inden-2-yl 4-methylbenzene-1-sulfonate
Synonyms: 2,3-dihydro-1H-inden-2-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 288.36
Molecular Formula: C16H16O3S
InChi Key: QUYCPYFNAZEWOT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1CC2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results