CAS RN 17797-10-3
IUPAC Name:
2-(4-fluorophenyl)propan-2-amine
Synonyms:
2-(4-fluorophenyl)propan-2-amine
Molecular Weight (g/mol):
153.2
Molecular Formula:
C9H12FN
InChi Key:
NYFSRXOWJXAHOO-UHFFFAOYSA-N
SMILES:
CC(C)(N)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
1-(4-Fluorophenyl)-1-methylethylamine, 97%
CAS: 17797-10-3 Molecular Formula: C9H12FN Molecular Weight (g/mol): 153.20 MDL Number: MFCD07777176 InChI Key: NYFSRXOWJXAHOO-UHFFFAOYSA-N Synonym: 2-4-fluorophenyl propan-2-amine,1-4-fluorophenyl-1-methylethylamine,1-4-fluorophenyl-1-methyl-ethylamine,1-4-fluoro-phenyl-1-methyl-ethylamine,2-4-fluorophenyl prop-2-ylamine,1-methyl-1-4-fluorophenyl ethylamine,4-fluorocumylamine,pubchem16914,4-fluoro-alpha,alpha-dimethylbenzylamine,alpha,alpha-dimethyl-4-fluorobenzylamine PubChem CID: 13025665 IUPAC Name: 2-(4-fluorophenyl)propan-2-amine SMILES: CC(C)(N)C1=CC=C(F)C=C1