accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17846-15-0

NH2HNO

CAS RN 17846-15-0

IUPAC Name: adamantane-1-carbohydrazide
Synonyms: adamantane-1-carbohydrazide
Molecular Weight (g/mol): 194.28
Molecular Formula: C11H18N2O
InChi Key: FJXQVRADKYKERU-UHFFFAOYSA-N
SMILES: NNC(=O)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results