accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NOOFFFF

CAS RN 179062-05-6

IUPAC Name: 4-fluoro-2-nitro-5-(trifluoromethyl)aniline
Synonyms: 4-fluoro-2-nitro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 224.12
Molecular Formula: C7H4F4N2O2
InChi Key: WOSGMZUIHAFPSS-UHFFFAOYSA-N
SMILES: NC1=C(C=C(F)C(=C1)C(F)(F)F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results