accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17933-03-8

IUPAC Name: (3-methylphenyl)boronic acid
Synonyms: 3-methylphenylboronic acid
Molecular Weight (g/mol): 135.96
Molecular Formula: C7H9BO2
InChi Key: BJQCPCFFYBKRLM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results