accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17938-13-5

CH3SiH3CCH3SiCH3H3CCH3

CAS RN 17938-13-5

IUPAC Name: trimethyl(2-{4-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)silane
Synonyms: trimethyl(2-{4-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)silane
Molecular Weight (g/mol): 270.52
Molecular Formula: C16H22Si2
InChi Key: CMTMWEXUJQSPCA-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results