accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 179420-75-8

CH3NNI

CAS RN 179420-75-8

IUPAC Name: 1-(4-iodophenyl)-2-methyl-1H-imidazole
Synonyms: 1-(4-iodophenyl)-2-methylimidazole
Molecular Weight (g/mol): 284.1
Molecular Formula: C10H9IN2
InChi Key: REVPKMONQSPHGF-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results