CAS RN 179474-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Prucalopride Succinate, TRC
CAS: 179474-85-2 Molecular Formula: C18 H26 Cl N3 O3 . C4 H6 O4 Molecular Weight (g/mol): 485.96 Synonym: Butanedioic acid, compd. with 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-7-benzofurancarboxamide (1:1),7-Benzofurancarboxamide, 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-, butanedioate (1:1) (9CI),Prucalopride succinate,R 108512,Resolor,Resotran IUPAC Name: 4-amino-5-chloro-N-[1-(3-methoxypropyl)piperidin-4-yl]-2,3-dihydro-1-benzofuran-7-carboxamide;butanedioic acid SMILES: COCCCN1CCC(CC1)NC(=O)c2cc(Cl)c(N)c3CCOc23.OC(=O)CCC(=O)O