accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1795-31-9

IUPAC Name: tritrimethylsilyl phosphite
Synonyms: tritrimethylsilyl phosphite
Molecular Weight (g/mol): 298.54
Molecular Formula: C9H27O3PSi3
InChi Key: VMZOBROUFBEGAR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)