accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHLu

CAS RN 17966-84-6

IUPAC Name: tris(4-hydroxypent-3-en-2-one) lutetium
Synonyms: tris(4-hydroxypent-3-en-2-one) lutetium
Molecular Weight (g/mol): 475.32
Molecular Formula: C15H24LuO6
InChi Key: BJUPKIHXMYXGRT-UHFFFAOYSA-N
SMILES: [Lu].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results