accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOONNHOCH3

CAS RN 179688-53-0

IUPAC Name: 7-methoxy-4-oxo-1,4-dihydroquinazolin-6-yl acetate
Synonyms: 7-methoxy-4-oxo-1H-quinazolin-6-yl acetate
Molecular Weight (g/mol): 234.21
Molecular Formula: C11H10N2O4
InChi Key: SOLQIFINSOHAQD-UHFFFAOYSA-N
SMILES: COC1=C(OC(C)=O)C=C2C(=O)N=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results