accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17969-22-1

IUPAC Name: 4-(chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole
Synonyms: 4-(chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole
Molecular Weight (g/mol): 244.13
Molecular Formula: C10H7Cl2NS
InChi Key: UEJQTBKTWJQBRN-UHFFFAOYSA-N
SMILES: ClCC1=CSC(=N1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results