accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1798-04-5

IUPAC Name: 2-(4-tert-butylphenoxy)acetic acid
Synonyms: 4-tert-butylphenoxyacetic acid
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: FBIGAJNVRFKBJL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(OCC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results