accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18000-24-3

OHOOClNH

CAS RN 18000-24-3

IUPAC Name: 7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
Synonyms: 7-chloro-kyurenic acid
Molecular Weight (g/mol): 223.61
Molecular Formula: C10H6ClNO3
InChi Key: UAWVRVFHMOSAPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)