accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1801-62-3

CH3NONSH3C

CAS RN 1801-62-3

IUPAC Name: 1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 144.19
Molecular Formula: C5H8N2OS
InChi Key: GKWNLFPHQCFFQJ-UHFFFAOYSA-N
SMILES: CN1CC(=O)N(C)C1=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results