accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1806-26-4

IUPAC Name: 4-octylphenol
Synonyms: 4-octylphenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: NTDQQZYCCIDJRK-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results