CAS RN 18063-03-1
IUPAC Name:
2,6-difluorobenzamide
Synonyms:
2,6-difluorobenzamide
Molecular Weight (g/mol):
157.12
Molecular Formula:
C7H5F2NO
InChi Key:
AVRQBXVUUXHRMY-UHFFFAOYSA-N
SMILES:
NC(=O)C1=C(F)C=CC=C1F
1
–
2
of
2
results
1
2,6-Difluorobenzamide, TRC
CAS: 18063-03-1 Molecular Formula: C7 H5 F2 N O Molecular Weight (g/mol): 157.12 Synonym: Benzamide, 2,6-difluoro-,2,6-Difluorobenzamide,Lufenuron Imp. A (EP) IUPAC Name: 2,6-difluorobenzamide SMILES: NC(=O)c1c(F)cccc1F
2
2,6-Difluorobenzamide, 98+%, Thermo Scientific™
CAS: 18063-03-1 Molecular Formula: C7H5F2NO Molecular Weight (g/mol): 157.12 MDL Number: MFCD00007972 InChI Key: AVRQBXVUUXHRMY-UHFFFAOYSA-N PubChem CID: 87439 IUPAC Name: 2,6-difluorobenzamide SMILES: C1=CC(=C(C(=C1)F)C(=O)N)F