accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNSOONOO

CAS RN 180631-81-6

IUPAC Name: N-(4-methoxyphenyl)-3-nitrobenzene-1-sulfonamide
Synonyms: N-(4-methoxyphenyl)-3-nitrobenzenesulfonamide
Molecular Weight (g/mol): 308.31
Molecular Formula: C13H12N2O5S
InChi Key: NHEAQAMNXXRBQT-UHFFFAOYSA-N
SMILES: COC1=CC=C(NS(=O)(=O)C2=CC=CC(=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results