accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18068-82-1

IUPAC Name: 2-phenylfuro[3,2-b]pyridine
Synonyms: 2-phenylfuro[3,2-b]pyridine
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: IZQVYGNVFNSARJ-UHFFFAOYSA-N
SMILES: O1C(=CC2=NC=CC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results