accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrH3C

CAS RN 1809-10-5

IUPAC Name: 3-bromopentane
Synonyms: pentane, 3-bromo-
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: VTOQFOCYBTVOJZ-UHFFFAOYSA-N
SMILES: CCC(Br)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results