accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1809-20-7

IUPAC Name: oxobis(propan-2-yloxy)-λ⁵-phosphanylium
Synonyms: diisopropoxy(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 165.15
Molecular Formula: C6H14O3P
InChi Key: BJLZAAWLLPMZQR-UHFFFAOYSA-N
SMILES: CC(C)O[P+](=O)OC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)