accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 181266-82-0

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHEu

CAS RN 181266-82-0

IUPAC Name: europium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Synonyms: europium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Molecular Weight (g/mol): 467.31
Molecular Formula: C15H23EuO7
InChi Key: GLDOQTGHEKXAIO-UHFFFAOYSA-K
SMILES: O.[Eu+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results